Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-3-methylisothiazole-5-carboxaldehyde 95+%
4-Bromo-3-methylisothiazole-5-carboxaldehyde 95+%
Catalog # APOSOR346491-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3-methylisothiazole-5-carboxaldehyde 95+%
Catalog # APOSOR346491-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.06 g/mol
Storage Temperature:Ambient
CAS Number:89283-96-5
Formula:C₅H₄BrNOS
MDL Number:MFCD27937097