1-Butyl-piperidine-4-carboxylic acid ethyl ester
Catalog # APOSOR346369-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.317 g/mol
Storage Temperature:Ambient
CAS Number:74045-89-9
Formula:C₁₂H₂₃NO₂
MDL Number:MFCD16152773