Order Entry
Czech Republic
ContactUsLinkComponent
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
Catalog # APOSOR346361-1G
Supplier:  Apollo Scientific
CAS Number:  
4,5,6,7-Tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
Catalog # APOSOR346361-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.177 g/mol
Storage Temperature:Ambient
CAS Number:307313-03-7
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD09056814