

6-Chloro-5-methyl-pyridine-3-sulphonyl chloride
Catalog # APOSOR346310-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:226.08 g/mol
Melting Pt:56…57 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
:GHS07
CAS Number:37105-10-5
Formula:C₆H₅Cl₂NO₂S
MDL Number:MFCD09881190