Order Entry
Czech Republic
ContactUsLinkComponent
4-Thiophen-3-yl-but-3-en-2-one
4-Thiophen-3-yl-but-3-en-2-one
Catalog # APOSOR346289-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Thiophen-3-yl-but-3-en-2-one
Catalog # APOSOR346289-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.214 g/mol
Storage Temperature:Ambient
CAS Number:123293-62-9
Formula:C₈H₈OS
MDL Number:MFCD20286588