Order Entry
Czech Republic
ContactUsLinkComponent
 
2-tert-Butoxycarbonylamino-4-methyl-oxazole-5-carboxylic acid methyl ester
Catalog # APOSOR346241-1G
Supplier:  Apollo Scientific
CAS Number:  
2-tert-Butoxycarbonylamino-4-methyl-oxazole-5-carboxylic acid methyl ester
Catalog # APOSOR346241-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.255 g/mol
Storage Temperature:Ambient
CAS Number:1311278-29-1
Formula:C₁₁H₁₆N₂O₅
MDL Number:MFCD19981434