2-Amino-5-methyl-nicotinonitrile
Catalog # APOSOR346129-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:133.151 g/mol
Storage Temperature:Ambient
CAS Number:38076-78-7
Formula:C₇H₇N₃
MDL Number:MFCD07779068