Order Entry
Czech Republic
ContactUsLinkComponent
1,2-Thiazinan-3-one 1,1-dioxide 97%
Irritant
1,2-Thiazinan-3-one 1,1-dioxide 97%
Catalog # APOSOR3458-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,2-Thiazinan-3-one 1,1-dioxide 97%
Catalog # APOSOR3458-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :149.168 g/mol
Melting Pt:150…152 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:913836-20-1
Formula:C₄H₇NO₃S
MDL Number:MFCD08689509