Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one
5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one
5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one
APOSOR345716-1G
:
Apollo Scientific
:
5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one
APOSOR345716-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
205.009 g/mol
:
Ambient
:
4722-76-3
:
C₅H₅BrN₂O₂
:
MFCD18427081