Order Entry
Czech Republic
ContactUsLinkComponent
4-Amino-2-methoxy-3-nitropyridine 98%
4-Amino-2-methoxy-3-nitropyridine 98%
Catalog # APOSOR345322-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-2-methoxy-3-nitropyridine 98%
Catalog # APOSOR345322-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :169.138 g/mol
Storage Temperature:Ambient
CAS Number:33623-16-4
Formula:C₆H₇N₃O₃
MDL Number:MFCD07437848