Order Entry
Czech Republic
ContactUsLinkComponent
2-Nitropyridine-3-carboxylic acid 97%
2-Nitropyridine-3-carboxylic acid 97%
Catalog # APOSOR345320-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Nitropyridine-3-carboxylic acid 97%
Catalog # APOSOR345320-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.107 g/mol
Storage Temperature:Ambient
CAS Number:33225-72-8
Formula:C₆H₄N₂O₄
MDL Number:MFCD04114130