2,4-Dibromo-3-hydroxy-6-methylpyridine 95+%
Catalog # APOSOR345302-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.918 g/mol
Storage Temperature:Ambient
CAS Number:23003-29-4
Formula:C₆H₅Br₂NO
MDL Number:MFCD09753265