(S)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340033-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:700.715 g/mol
Melting Pt:265…278 °C
Storage Temperature:Ambient
CAS Number:361342-52-1
Formula:C₄₈H₂₉O₄P
MDL Number:MFCD11045406