
(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340031-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:590.476 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:878111-16-1
Formula:C₃₂H₁₉N₂O₈P
MDL Number:MFCD08274951