Order Entry
Czech Republic
ContactUsLinkComponent
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Irritant
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340024-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSOR340024-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :752.959 g/mol
Melting Pt:210…213 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:791616-63-2
Formula:C₅₀H₅₇O₄P
MDL Number:MFCD10567009