Order Entry
Czech Republic
ContactUsLinkComponent
5-Methyl-1H,2H,7H-pyrido[3,2-d][1,2]diazepin-2-one, tech
5-Methyl-1H,2H,7H-pyrido[3,2-d][1,2]diazepin-2-one, tech
Catalog # APOSOR33566-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methyl-1H,2H,7H-pyrido[3,2-d][1,2]diazepin-2-one, tech
Catalog # APOSOR33566-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.187 g/mol
Storage Temperature:Ambient
CAS Number:1092344-97-2
Formula:C₉H₉N₃O
MDL Number:MFCD11501765