Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine-6-carboxylate, tech
Methyl 3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine-6-carboxylate, tech
Catalog # APOSOR33431-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-[(dimethylamino)methyl]imidazo[1,2-a]pyridine-6-carboxylate, tech
Catalog # APOSOR33431-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.266 g/mol
Storage Temperature:Ambient
CAS Number:882748-14-3
Formula:C₁₂H₁₅N₃O₂
MDL Number:MFCD08064773