Order Entry
Czech Republic
ContactUsLinkComponent
2-(5-Oxo-4-phenyl-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetonitrile, tech
2-(5-Oxo-4-phenyl-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetonitrile, tech
Catalog # APOSOR33427-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(5-Oxo-4-phenyl-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetonitrile, tech
Catalog # APOSOR33427-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.197 g/mol
Storage Temperature:Ambient
CAS Number:866149-44-2
Formula:C₁₀H₈N₄O
MDL Number:MFCD04126418