Order Entry
Czech Republic
ContactUsLinkComponent
 
2-[4-(4-Methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Catalog # APOSOR33425-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[4-(4-Methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Catalog # APOSOR33425-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.223 g/mol
Storage Temperature:Ambient
CAS Number:866149-37-3
Formula:C₁₁H₁₀N₄O
MDL Number:MFCD04126410