Ethyl 2,5,7-trimethylimidazo[1,2-a]pyridine-3-carboxylate, tech
Catalog # APOSOR33375-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.278 g/mol
Storage Temperature:Ambient
CAS Number:81438-54-2
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD02082626