Order Entry
Czech Republic
ContactUsLinkComponent
6-(4-Chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
6-(4-Chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR33354-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(4-Chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR33354-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.676 g/mol
Storage Temperature:Ambient
CAS Number:339109-70-5
Formula:C₁₃H₉ClN₂O
MDL Number:MFCD00202815