(E)-N,N-Dimethyl-N'-(5-methyl-1,2-oxazol-3-yl)methanimidamide 95%
Catalog # APOSOR33342-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:153.182 g/mol
Storage Temperature:Ambient
CAS Number:134540-15-1
Formula:C₇H₁₁N₃O
MDL Number:MFCD00117221