(2E)-3-(Dimethylamino)-1-[4-(4-methylpiperazin-1-yl)phenyl]prop-2-en-1-one, tech
Catalog # APOSOR33323-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:273.373 g/mol
Storage Temperature:Ambient
CAS Number:338975-38-5
Formula:C₁₆H₂₃N₃O
MDL Number:MFCD00138783