Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Methoxy-1-benzofuran-2-carbonitrile
7-Methoxy-1-benzofuran-2-carbonitrile, tech
7-Methoxy-1-benzofuran-2-carbonitrile, tech
7-Methoxy-1-benzofuran-2-carbonitrile, tech
APOSOR33265-1G
:
Apollo Scientific
:
7-Methoxy-1-benzofuran-2-carbonitrile, tech
APOSOR33265-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
173.168 g/mol
:
Ambient
:
35351-49-6
:
C₁₀H₇NO₂
:
MFCD02082422