Order Entry
Czech Republic
ContactUsLinkComponent
3-Benzyl-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech
3-Benzyl-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR33262-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Benzyl-4-hydroxy-1-(2-methoxyethyl)-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR33262-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.3 g/mol
Storage Temperature:Ambient
CAS Number:339106-88-6
Formula:C₁₅H₁₇NO₃
MDL Number:MFCD01568818