Order Entry
Czech Republic
ContactUsLinkComponent
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-3,3-dimethylazetidin-2-one, tech
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-3,3-dimethylazetidin-2-one, tech
Katalogové číslo APOSOR33242-1G
Supplier:  Apollo Scientific
CAS Number:  
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-3,3-dimethylazetidin-2-one, tech
Katalogové číslo APOSOR33242-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.342 g/mol
Storage Temperature:Ambient
CAS Number:339100-33-3
Formula:C₁₆H₂₀N₂O₂
MDL Number:MFCD00793419