Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Dimethyl-5-(prop-2-en-1-yloxy)-1H-pyrazole-4-carbaldehyde 95%
1,3-Dimethyl-5-(prop-2-en-1-yloxy)-1H-pyrazole-4-carbaldehyde 95%
Catalog # APOSOR33228-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Dimethyl-5-(prop-2-en-1-yloxy)-1H-pyrazole-4-carbaldehyde 95%
Catalog # APOSOR33228-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.204 g/mol
Storage Temperature:Ambient
CAS Number:321574-23-6
Formula:C₉H₁₂N₂O₂
MDL Number:MFCD00665212