Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile 95%
2-(2-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile 95%
Catalog # APOSOR33226-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile 95%
Catalog # APOSOR33226-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.697 g/mol
Storage Temperature:Ambient
CAS Number:66549-12-0
Formula:C₁₂H₁₃ClN₂O
MDL Number:MFCD00173632