3-(4-Chlorophenoxy)butan-2-one, tech
Catalog # APOSOR33209-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.646 g/mol
Storage Temperature:Ambient
CAS Number:3782-11-4
Formula:C₁₀H₁₁ClO₂
MDL Number:MFCD00243668