Order Entry
Czech Republic
ContactUsLinkComponent
1-[2-(4-Chlorophenyl)-2-oxoethyl]-1,2-dihydropyridin-2-one, tech
1-[2-(4-Chlorophenyl)-2-oxoethyl]-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR33208-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(4-Chlorophenyl)-2-oxoethyl]-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR33208-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.677 g/mol
Storage Temperature:Ambient
CAS Number:108664-55-7
Formula:C₁₃H₁₀ClNO₂
MDL Number:MFCD00139074