2,3,4,5-Tetrahydro-1H-1,5-benzodiazepin-2-one 95%
Catalog # APOSOR33169-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.189 g/mol
Storage Temperature:Ambient
CAS Number:5755-07-7
Formula:C₉H₁₀N₂O
MDL Number:MFCD00612448