Order Entry
Czech Republic
ContactUsLinkComponent
4,6-Dimethyl-2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
4,6-Dimethyl-2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR33152-1G
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dimethyl-2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR33152-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.226 g/mol
Storage Temperature:Ambient
CAS Number:675830-24-7
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD03217148