Order Entry
Czech Republic
ContactUsLinkComponent
N-(2-Chloroethyl)-1H,2H-naphtho[2,1-b]furan-2-carboxamide, tech
N-(2-Chloroethyl)-1H,2H-naphtho[2,1-b]furan-2-carboxamide, tech
Catalog # APOSOR33145-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(2-Chloroethyl)-1H,2H-naphtho[2,1-b]furan-2-carboxamide, tech
Catalog # APOSOR33145-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.73 g/mol
Storage Temperature:Ambient
CAS Number:478078-36-3
Formula:C₁₅H₁₄ClNO₂
MDL Number:MFCD02570920