3-(4-Methoxyphenyl)-1-(pyridin-2-yl)propan-1-ol, tech
Catalog # APOSOR33144-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.301 g/mol
Storage Temperature:Ambient
CAS Number:478078-18-1
Formula:C₁₅H₁₇NO₂
MDL Number:MFCD02570997