Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 2-(4,8-dimethyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech
Ethyl 2-(4,8-dimethyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech
Catalog # APOSOR33133-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-(4,8-dimethyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech
Catalog # APOSOR33133-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.3 g/mol
Storage Temperature:Ambient
CAS Number:145667-25-0
Formula:C₁₅H₁₇NO₃
MDL Number:MFCD01444200