(2E)-3-(Dimethylamino)-1-[4-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one, tech
Catalog # APOSOR33131-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.299 g/mol
Storage Temperature:Ambient
CAS Number:339018-66-5
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD01443774