Order Entry
Czech Republic
ContactUsLinkComponent
3-(4-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol, tech
3-(4-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol, tech
Catalog # APOSOR33115-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol, tech
Catalog # APOSOR33115-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.643 g/mol
Storage Temperature:Ambient
CAS Number:126542-93-6
Formula:C₁₀H₈ClN₃O
MDL Number:MFCD05668805