Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Acetylphenyl)-3,3-dimethylazetidin-2-one, tech
1-(4-Acetylphenyl)-3,3-dimethylazetidin-2-one, tech
Catalog # APOSOR33107-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Acetylphenyl)-3,3-dimethylazetidin-2-one, tech
Catalog # APOSOR33107-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.264 g/mol
Storage Temperature:Ambient
CAS Number:339015-93-9
Formula:C₁₃H₁₅NO₂
MDL Number:MFCD00793414