2-Methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile 95%
Catalog # APOSOR33076-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:158.16 g/mol
Storage Temperature:Ambient
CAS Number:339010-67-2
Formula:C₈H₆N₄
MDL Number:MFCD00173619