Order Entry
Czech Republic
ContactUsLinkComponent
1-[(4-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid, tech
1-[(4-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid, tech
Catalog # APOSOR33071-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(4-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid, tech
Catalog # APOSOR33071-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :263.676 g/mol
Storage Temperature:Ambient
CAS Number:339008-74-1
Formula:C₁₃H₁₀ClNO₃
MDL Number:MFCD00139926