Order Entry
Czech Republic
ContactUsLinkComponent
(3Z)-3-{[(2-Hydroxyphenyl)amino]methylidene}-1,3-dihydro-2-benzofuran-1-one, tech
(3Z)-3-{[(2-Hydroxyphenyl)amino]methylidene}-1,3-dihydro-2-benzofuran-1-one, tech
Catalog # APOSOR33069-1G
Supplier:  Apollo Scientific
CAS Number:  
(3Z)-3-{[(2-Hydroxyphenyl)amino]methylidene}-1,3-dihydro-2-benzofuran-1-one, tech
Catalog # APOSOR33069-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.253 g/mol
Storage Temperature:Ambient
CAS Number:339008-47-8
Formula:C₁₅H₁₁NO₃
MDL Number:MFCD00139664