Order Entry
Czech Republic
ContactUsLinkComponent
1-(Morpholin-4-yl)-3-phenoxypropan-2-ol 95%
1-(Morpholin-4-yl)-3-phenoxypropan-2-ol 95%
Catalog # APOSOR33045-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(Morpholin-4-yl)-3-phenoxypropan-2-ol 95%
Catalog # APOSOR33045-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.295 g/mol
Storage Temperature:Ambient
CAS Number:5296-26-4
Formula:C₁₃H₁₉NO₃
MDL Number:MFCD00450856