Order Entry
Czech Republic
ContactUsLinkComponent
5,5-Dimethyl-2-[(phenylamino)methylidene]cyclohexane-1,3-dione, tech
5,5-Dimethyl-2-[(phenylamino)methylidene]cyclohexane-1,3-dione, tech
Catalog # APOSOR33041-1G
Supplier:  Apollo Scientific
CAS Number:  
5,5-Dimethyl-2-[(phenylamino)methylidene]cyclohexane-1,3-dione, tech
Catalog # APOSOR33041-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.301 g/mol
Storage Temperature:Ambient
CAS Number:29974-49-0
Formula:C₁₅H₁₇NO₂
MDL Number:MFCD00512921