Order Entry
Czech Republic
ContactUsLinkComponent
5,5-Dimethyl-2-{[(pyridin-3-yl)amino]methylidene}cyclohexane-1,3-dione, tech
5,5-Dimethyl-2-{[(pyridin-3-yl)amino]methylidene}cyclohexane-1,3-dione, tech
Catalog # APOSOR33040-1G
Supplier:  Apollo Scientific
CAS Number:  
5,5-Dimethyl-2-{[(pyridin-3-yl)amino]methylidene}cyclohexane-1,3-dione, tech
Catalog # APOSOR33040-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.289 g/mol
Storage Temperature:Ambient
CAS Number:419542-34-0
Formula:C₁₄H₁₆N₂O₂
MDL Number:MFCD01310105