Order Entry
Czech Republic
ContactUsLinkComponent
5-(2,3-Dihydro-1H-inden-5-yl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
5-(2,3-Dihydro-1H-inden-5-yl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
Catalog # APOSOR33037-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(2,3-Dihydro-1H-inden-5-yl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
Catalog # APOSOR33037-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.293 g/mol
Storage Temperature:Ambient
CAS Number:866050-30-8
Formula:C₁₆H₁₂N₄
MDL Number:MFCD04125854