Order Entry
Czech Republic
ContactUsLinkComponent
1-[6-(4-Methoxyphenoxy)-2-methylpyridin-3-yl]ethan-1-one, tech
1-[6-(4-Methoxyphenoxy)-2-methylpyridin-3-yl]ethan-1-one, tech
Catalog # APOSOR33026-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[6-(4-Methoxyphenoxy)-2-methylpyridin-3-yl]ethan-1-one, tech
Catalog # APOSOR33026-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.284 g/mol
Storage Temperature:Ambient
CAS Number:478064-80-1
Formula:C₁₅H₁₅NO₃
MDL Number:MFCD02570895