N'-(6-Chloropyridin-2-yl)-2,2-dimethylpropanehydrazide, tech
Catalog # APOSOR33013-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.691 g/mol
Storage Temperature:Ambient
CAS Number:66999-53-9
Formula:C₁₀H₁₄ClN₃O
MDL Number:MFCD02187190