1-(4-Methyl-2-phenyl-1,3-thiazol-5-yl)ethan-1-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7520-94-7
:
Formula:C₁₂H₁₁NOS
MDL Number:MFCD01871613
:217.287 g/mol
Storage Temperature:Ambient
Frequently Bought Together