Order Entry
Czech Republic
ContactUsLinkComponent
5-Amino-6-methyl-4H,5H-furo[3,2-c]pyridin-4-one, tech
5-Amino-6-methyl-4H,5H-furo[3,2-c]pyridin-4-one, tech
Catalog # APOSOR33006-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Amino-6-methyl-4H,5H-furo[3,2-c]pyridin-4-one, tech
Catalog # APOSOR33006-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.161 g/mol
Storage Temperature:Ambient
CAS Number:57053-27-7
Formula:C₈H₈N₂O₂
MDL Number:MFCD01443847