Order Entry
Czech Republic
ContactUsLinkComponent
 
3-Chloro-N-(4-methoxyphenyl)-2,2-dimethylpropanamide, tech
Catalog # APOSOR32992-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-N-(4-methoxyphenyl)-2,2-dimethylpropanamide, tech
Catalog # APOSOR32992-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.714 g/mol
Storage Temperature:Ambient
CAS Number:338968-17-5
Formula:C₁₂H₁₆ClNO₂
MDL Number:MFCD00793409